High Five™ SMS Disposable Lab Coats

Breathable for all day comfort

Manufacturer: High Five™ AL454

 View more versions of this product

Catalog No. 18-156-200B

  Check Availability
Add to Cart

    Specifications

    Material SMS (Spunbound Melt-Blown Synthetic)
    Size X-Large
    For Use With (Application) Protects from dust, dirt, grime and Water based liquids
    Closure Type Snap
    Cuff Style Knit
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit