Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High Five™ SMS Disposable Lab Coats
Click to view available options
Color:
Blue
Purple
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
Description
Breathable for all day comfort
- Protect against dry particulates and water-based liquids
- Knit collar and cuffs
- Five-snap front closure
- Available in white or blue
Specifications
Size | Small |
Material | SMS |
Color | Blue |
Closure Type | Snaps |
No. of Pockets | 3 |
For Use With (Application) | Protects from dust, dirt, grime and Water based liquids |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction