Learn More
High Five™ SMS Disposable Lab Coats Get IT Item
Breathable for all day comfort
Manufacturer:  High Five™ EB152043XL
 View more versions of this product
Catalog No. 19-167-633
Add to cart
Description
Description
Breathable for all day comfort
  • Protect against dry particulates and water-based liquids
  • Knit collar and cuffs
  • Five-snap front closure
  • Available in white or blue
Specifications
Specifications
Snap
SMS (Spunbound Melt-Blown Synthetic)
3X-Large
Blue
3
Protects from dust, dirt, grime and Water based liquids