Learn More
High Five™ SMS Isolation Gowns
Gives added durability
Manufacturer:  High Five™ AG304
Catalog No. 19-167-919
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Wraparound design shields clothing from particulates and light splash
  • 3-layer construction offers durability with comfort due to its breathability
  • Neck and waist ties
  • Color: Blue
Specifications
Specifications
For long durability, protects against particulate and light splash
Blue
X-Large
3-ply
SMS