Learn More
High Five™ Spunbond Isolation Gowns
Durable protection from common dirt, dust and grime, minor splashes and particulates
Manufacturer:  High Five™ EY10214M
 View more versions of this product
Catalog No. 19-168-665
Add to cart
Description
Description
Spunbond polypropylene material protects arms, chest, shoulders, waist and upper legs from common workplace particulates.
  • Lightweight, breathable fabric keeps workers cool and comfortable
  • Elastic wrists with easy neck and waist ties for wrap-around styling
Specifications
Specifications
Protection from common dirt, dust and grime, minor splashes and particulates
Yellow
Medium
Elastic
Spunbond polypropylene