High Five™ Spunbond Polyproplyene Shoe Cover

Manufacturer: High Five™ AS693

 View more versions of this product

Catalog No. 19-156-244

  Check Availability
Add to Cart

    Specifications

    Material Spunbond Polypropylene
    Closure Type Elastic
    Color White
    Size Universal
    Product Type Shoe Covers
    For Use With (Application) Ideal for protection against dirt, debris and light splashes
    Includes Tread, 2.5 in. wide tacky strip

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit