Learn More
High Five™ Spunbond Polypropylene Coveralls
Click to view available options
Cuff Style:
Elastic
Knit
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Spunbond polypropylene
- Porous construction for breathability
- Elastic wrists and ankles
- Zipper closure
- White
Specifications
Size | 3X-Large |
Material | Polypropylene |
Color | White |
Ankle Style | Elastic |
Cuff Style | Elastic |