High Five™ Spunbond Polypropylene Lab Coats

Economical protection from dust, dirt and grime

Manufacturer: High Five™ AL616

 View more versions of this product

Catalog No. 19-167-075

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Material Spunbond Polypropylene
    No. of Pockets 3.0
    Size 3X-Large
    For Use With (Application) Protection from dust, dirt and grime
    Closure Type Snap Front
    Color Clear
    Construction Breathable Construction Keeps Workers Cooler

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit