Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High Five™ Spunbond Polypropylene Lab Coats
Click to view available options
Color:
Navy
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
Economical protection from dust, dirt and grime
- Breathable construction keeps workers cooler
- No pockets to catch contaminants
- Elastic wrists
- Four-snap closure
- White
Specifications
Size | Large |
Material | Polypropylene |
Color | White |
Closure Type | Snaps |
For Use With (Application) | Protection from dust, dirt and grime |
Construction | Breathable Construction Keeps Workers Cooler |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction