High Five™ Spunbond Polypropylene Shoe Covers with Tread

Sanitary, with 2.5 in. tacky strip on sole to keep wearer from slipping

Manufacturer: High Five™ AS623

 View more versions of this product

Catalog No. 19-167-941

  Check Availability
Add to Cart

    Specifications

    Size Universal
    Product Type Shoe Covers
    Material Spunbond Polypropylene
    For Use With (Application) Ideal to provide protection against general splash and spills
    Closure Type Elastic
    Packaging Type Bulk
    Color Blue
    Includes Blue and white Tread, 2.5 in. W tacky strip

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit