High Five™ Velcro™ PPSB Lab Coats

Velcro closure for an adjustable fit

Manufacturer: High Five™ AL706

 View more versions of this product

Catalog No. 19-156-166

  Check Availability
Add to Cart

    Specifications

    Material SMS (Spunbound Melt-Blown Synthetic)
    Item Description Velcro™
    Closure Type Hook and Loop
    Construction Breathable and Comfortable Spunbond Polypropylene
    Size 3X-Large
    For Use With (Application) Protection from dust, dirt and grime
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit