Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High Five™ PPSB Lab Coats
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Breathable and comfortable spunbound polypropylene
- Protects from dust and grime
- Closure stays securely shut
- White
Specifications
Size | Small |
Material | Polypropylene |
Color | White |
Closure Type | Hook and Loop |
For Use With (Application) | Protection from dust, dirt and grime |
Construction | Breathable and Comfortable Spunbond Polypropylene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction