Learn More
Amerex™ High-Performance Fire Extinguisher with ABC Compliance Flow
Amerex™ High-Performance Fire Extinguishers are available with a proprietary, zinc-enriched coating that resists corrosion for up to 12 years in severely salt-contaminated environments.
Supplier:  Amerex™ 26912
 View more versions of this product
Catalog No. 19-860-558
Add to Cart
Description
Description
  • Features zinc enriched primer coating
  • Resists extreme environmental conditions
  • Highest achievable UL ratings
  • Proprietary corrosion-resistant paint process
  • Simple operation and maintenance
  • Improved environmental impact compared to galvanized units
  • ABC Compliance Flow
Specifications
Specifications
Fire Extinguisher
Stainless Steel, Brass, Steel
6 kg
22 sec.
713
4A:80B:C
14 kg
Red
13.2 lb.
7 x 10.5 x 17.5 in. (17.78 x 26.67 x 44.5 cm)
Wall Hanger, Protective Foot Ring
40 to 50 ft.
31 lb.