Learn More
Amerex™ High-Performance Fire Extinguisher with ABC Fast Flow
Amerex™ High-Performance Fire Extinguishers are available with a proprietary, zinc-enriched coating that resists corrosion for up to 12 years in severely salt-contaminated environments.
Supplier:  Amerex™ 26904
 View more versions of this product
Catalog No. 19-860-567
Add to Cart
Description
Description
  • Features zinc enriched primer coating
  • Resists extreme environmental conditions
  • Highest achievable UL ratings
  • Proprietary corrosion-resistant paint process
  • Simple operation and maintenance
  • Improved environmental impact compared to galvanized units
  • ABC Fast Flow
Specifications
Specifications
Fire Extinguisher
Stainless Steel, Brass, Steel
9 kg
18 sec.
791
4A:40B:C
19.5 kg
Red
20 lb.
7 x 10.5 x 24.75 in. (17.78 x 26.67 x 62.87 cm)
Wall Hanger, Protective Foot Ring
40 to 50 ft.
43 lb.