Learn More
Brady™ High Performance Pipe Marker for Water - Coiled Printed Plastic, Style II, COLD WATER RETURN
Style II wrap-around pipe markers fit pipes with outside Dia. of 2-1/2 in. to 7-7/8 in.
Supplier:  Brady™ 80550
Catalog No. 19-112-333
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Coiled Printed Plastic, Style II, COLD WATER RETURN
Specifications
Specifications
Not Autoclavable
Pipe Marker
White on Green
Pipe Marker
1 Ea.
12 x 26.5 in.
12 in.
ANSI A13.1-2007
Non-adhesive
Polyester
English
26.5 x 12 in. (W x H)
26.5 in.
Product Suggestions
Product Suggestions