Learn More
ML Kishigo High Performance Surveyors Vest
ML Kishigo High Performance Surveyors Vest with ANSI 107 Class 3 Safety Compliance
Manufacturer:  ML Kishigo S50142XL
 View more versions of this product
Catalog No. 19-441-096
Add to cart
Description
Description
  • 100% polyester mesh material
  • Zipper front closure reinforced with durable webbing
  • 2 in. wide high performance reflective material with 2.5 in. contrasting black
  • Padded neck to ensure comfort even when vest is weighed down with tools
  • Left and right mic tabs
  • Two lapel grommets
Specifications
Specifications
ANSI 107 Class 3
Lime
Polyester
74 cm
Mesh
2X