Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan™ High Pressure Syringe
Designed for high pressure gas sampling
Supplier: Trajan™ 008170
Description
- Designed for high pressure gas sampling (rated to 500psi)
- Fitted with a gas tight ON/OFF valve and high pressure 7cm, 23 gauge needle
Specifications
23 G | |
0.63 mm | |
1/Ea. | |
Bevel |
0.37 mm | |
High Pressure Syringe | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title