Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ High-Select™ Phosphopeptide Enrichment Kit

Enriches phosphopeptides from complex samples.

$373.36 - $515.32

Specifications

Kit Contents Spin columns, white luer plug (end cap), binding/wash buffer, phosphopeptide elution buffer
Storage 4°C
Detection Mass spectrometry
Product Type Enrichment kit
Format 30 reactions
View More Specs

Products 2
Catalog Number Mfr. No. Description Format Price Quantity & Availability  
Catalog Number Mfr. No. Description Format Price Quantity & Availability  
PIA32993 View Documents Thermo Scientific™
A32993
Enriches TiO2 phosphopeptide from complex samples 24 reactions Each for $515.32
Add to cart
 
PIA32992 View Documents Thermo Scientific™
A32992
Enriches Fe-NTA phosphopeptide from complex samples 30 reactions Each for $373.36
Add to cart
 
Description

Description

  • Enriches phosphopeptides from complex samples using iron-chelate resin in spin columns
  • Starting sample: 0.5 to 5mg (of total protein digest)
  • Enrichment upto 150µg of phosphopeptides
    Convenient pre-formulated buffers for ease-of-use and spin-column format to enable parallel processing of multiple samples
  • High binding capacity
  • High specificity recovers phosphopeptides with >90% selectivity
  • High coverage recovers peptides with single and multiple phosphorylation sites
Specifications

Specifications

Spin columns, white luer plug (end cap), binding/wash buffer, phosphopeptide elution buffer
Mass spectrometry
30 reactions
Microcentrifuge
4°C
Enrichment kit
Mass spectrometry, enrichment of phosphorylated peptides
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit