Learn More
METTLER TOLEDO™ High Temp Differential Scanning Calorimeter (DSC)
Measure samples with the most sensitive Differential Scanning Calorimeter on the market – ideal for investigating all kinds of materials and effects. Our revolutionary star-shaped arrangement of thermocouples guarantees flat baselines and reproducible measurement results.
Manufacturer:  METTLER TOLEDO™ 30421564
 View more versions of this product
Catalog No. 01-915-189
Add to cart
Description
Description
  • Rugged MultiSTARe sensor with 56 thermocouples – detects the smallest and largest thermal effects
  • Robust endurance-tested sample robot – operates efficiently and reliably around the clock
  • Outstanding resolution – allows measurement of rapid changes and close-lying effects
  • Small and large sample volumes – for microgram or inhomogeneous samples
  • Start the experiment with just One Click™ – fast and easy routine operation
  • Simple FlexCal™calibration – saves time and results in precise and accurate measurements
  • Modular concept protects your investment – fulfills your current and future needs
  • Wide temperature range – from –150 to 700 °C in one measurement
  • User replaceable sensor – Separate sensor and furnace allow for cost effective maintenance
  • Corrosion resistant materials – Gold and palladium thermocouples embedded within a ceramic sensor prevent oxidation
  • Intelligent ergonomic design – simplifies instrument operation
Specifications
Specifications
IEC/EN61010-1:2001
24.8 in.
±0.02K
115V
High Temp Premium DSC with Sample Robot
24.8 in.
50/60Hz
RT to 700°C
18.1 in.
Robot