Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals High Unsaturated Fat Diet, Pellet

High in safflower oil. High in unsaturated fats. 4.39 kcal/gm.

$643.50 - $2420.00

Specifications

Type Animal Diet
Storage Requirements Refrigerated
Disposable Yes
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP296024410
View Documents
MP Biomedicals
0296024410
10 kg
Each for $643.50
Add to cart
 
MP296024420
View Documents
MP Biomedicals
0296024420
20 kg
Each for $1,114.00
Add to cart
 
MP296024450
View Documents
MP Biomedicals
0296024450
50 kg
Each for $2,420.00
Add to cart
 
Description

Description

  • High in Safflower oil
  • High in unsaturated fats
  • 4.39 kcal/gm
  • Contains casein, methionine, corn starch, mineral and vitamin fortification mix
Specifications

Specifications

Animal Diet
Refrigerated
Yes
Non-sterile
SDS
Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit