Learn More
Synthware™ High Vacuum Valve Plug, PTFE Protected
Complete with control knob. Viton™ O-rings on PTEF shaft are protected with precision PTFE wipers.
Manufacturer:  Synthware™ S600004
 View more versions of this product
Catalog No. 31-502-065
Add to cart
Description
Description
This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.
Specifications
Specifications
Organic Synthesis
Borosilicate Glass
0 to 4 mm High Vacuum Valve