Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hikeshi Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148081AP

Catalog No. 50-172-9738


Add to cart

Description

Description

Required for organization and/or function of the secretory apparatus in Clara cells in lung.
Specifications

Specifications

Hikeshi
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q53FT3, Q5M808, Q9DD02
C11orf73, Hikeshi, HSPC138, HSPC179, HSPC248, L7RN6
Rabbit
IgG
150 μL
-20°C
Primary
293103, 51501, 67669
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HIKESHI
Liquid
HIKESHI
Hikeshi Fusion Protein Ag6530
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit