Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ HILED Lighting Option
GREENER_CHOICESDP

Catalog No. 01257238 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Color:
Green, Blue, Far Red
Green, Blue, Infa Red, Far Red
Infa Red, Far Red
Red, Blue, Green
Red, Blue, Green, InfraRed
Red, InfraRed
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-257-238 Red, Blue, Green, InfraRed
01-257-237 Red, Blue, Green
01-257-242 Green, Blue, Far Red
01-257-243 Green, Blue, Infa Red, Far Red
01-257-240 Red, InfraRed
01-257-239 Infa Red, Far Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-257-238 Supplier Syngene™ Supplier No. GMHILEDRGBIR
Add to Cart
Add to Cart
Request a Quote

HILED lighting option for use with GBOX Mini includes gantry, electronics, and lighting units.

  • For use with GBOX Mini only
  • Including gantry, electronics, and lighting units

Specifications

Product Type Lighting
Color Red, Blue, Green, InfraRed
For Use With (Equipment) GBOX Mini
Includes Gantry, electronics, lighting units
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.