Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syngene™ HILED Upgrade Lighting from RGB to RGBIR



Click to view available options
Color:
Far Red
Infra Red
Red, Green, Blue, UV
Description
For use with GBOX
Specifications
Specifications
For Use With (Application) | Multiplex Western Fluorescence |
Color | Red, Green, Blue, UV |
For Use With (Equipment) | Gbox Chemi Systems |
Quantity | 1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction