Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare HiLoad™ 16/600 Superdex™ 200 pg: PROMO

Use for separating MAbs from dimers and contaminants. GE Healthcare HiLoad™ 16/600 Superdex™ 200 pg is ideal for high-resolution preparative gel filtration chromatography.

Manufacturer:  GE Healthcare 28989335PM

Catalog No. 45-002-490LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Includes:
  • Dismantling tool
  • Support screen
  • Net ring (10μm)
  • O-ring
  • Stop plugs
  • Instructions

Description

Description

Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose. Dextran component determined the separation properties of these media. Steep selectivity curves give gppd resolving power for peptides and proteins in the molecular weight range Mr 10 000 600 000 (Superdex 200 pg).

  • Used for separating MAbs from dimers and contaminants
  • Short run times with high recovery
  • Medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions
  • Separations can be scaled up to production levels
  • 0.06 in. fittings to connect to AKTA design systems
  • Theoretical plates: >13000m-1
  • Flow rate: <1.6mL/min.
  • Pressure max. (over the packed bed during operation): 3 bar [0.3MPa] (42psi)

  • Media
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH Stability working range: 3 to 12
  • pH Stability cleaning range: 2 to 14
  • Fractionation [Mr] globular proteins: 1 × 104-6 × 105
  • Fractionation [Mr] dextrans: 1 × 103-1 × 105
  • Shelf life from manufacture date: 5 years
  • Chemical stability: all commonly used buffers, 1M NaOH, 0.1M HCl, 1M acetic acid, 8M urea, 6M guanidine HCl, 1% SDS, 70% ethanol, 30% iso-propanol, 30% acetonitrile
  • Column
  • Column I.D.: 0.62 in. (16mm)
  • Inlet I.D.: 0.031 in. (0.8mm)
  • Outlet I.D.: 0.031 in. (0.8mm)
  • Chemical stability: resistant to aqueous solutions and solvents used in liquid chromatography. Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol
  • Column hardware made from borosilicate glass, polyamide, fuororubber, Ethylene Propylene Diene (EPDM), ETFE, Superpolyoxymethylene
  • Column tube made from borosilicate glass
  • Bed support made from Polypropylene (PP) (reinforced glass fiber), Polyamide (PA)
  • Porosity: 10μm

Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids

Order Info

  • HiLoad columns are prepacked XK columns
  • Bed volume: 120mL
  • Bed dimensions: 0.62 x 23.62 in. (16 × 600mm)
  • Sample volume: 5mL

Specifications

Specifications

<5mL
4 to 30°C, 20% Ethanol
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit