Learn More
HIN-1/SCGB3A1 Goat anti-Mouse, Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF2954SP
Catalog No. AF2954SP
Add to cart
This item is not returnable. View return policy
Description
Description
HIN-1/SCGB3A1 Polyclonal antibody AF2954SP specifically detects HIN-1/SCGB3A1 in Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
HIN-1/SCGB3A1
LYOPH
Western Blot 1 ug/mL
Q920D7
cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse HIN-1/SCGB3A1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human HIN-1 is observed.
Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Otud4
E. coli-derived recombinant mouse HIN-1/SCGB3A1 Phe22-Gly104 Accession # Q920D7
Immunogen affinity purified
RUO
Primary
92304
Reconstitute at 0.2 mg/mL in sterile PBS.