Learn More
Elma™ Hinged Lids for Elmasonic Xtra ST Ultrasonic Cleaner
For use with Elmasonic Xtra ST Ultrasonic Cleaner.
Supplier: Elma™ 107 6206
Description
- Stainless steel construction
- Helps to reduce noise and evaporation, serves as drip trays
- Insulated for sound reduction
Specifications
Lid | |
Hinged Lid | |
Stainless Steel |
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H | |
Helps to Reduce Noise and Evaporation, Serves as Drip Trays |