Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HINT2 Mouse anti-Human, Mouse, Clone: 5B10, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP25938750UL
Description
HINT2 Monoclonal specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA.Specifications
HINT2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2 | |
Mouse | |
Protein A purified | |
RUO | |
84681 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
5B10 | |
Unconjugated | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
HINT2 | |
Recombinant human HINT2 (18-163aa) purified from E. coli | |
50 μL | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only