Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HINT2 Mouse anti-Human, Mouse, Clone: 5B10, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP259387100UL

 View more versions of this product

Catalog No. NBP259387X



Description

Description

HINT2 Monoclonal specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

HINT2
Monoclonal
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2
Mouse
Protein A purified
RUO
84681
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
5B10
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
HINT2
Recombinant human HINT2 (18-163aa) purified from E. coli
100 μL
Primary
Human, Mouse
Purified
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only