Learn More
HINT2 Mouse anti-Human, Mouse, Clone: 5B10, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$300.50 - $458.00
Specifications
Antigen | HINT2 |
---|---|
Clone | 5B10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25938750
|
Novus Biologicals
NBP25938750UL |
50 μL |
Each for $300.50
|
|
|||||
NBP259387X
|
Novus Biologicals
NBP259387100UL |
100 μL |
Each for $458.00
|
|
|||||
Description
HINT2 Monoclonal specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA.Specifications
HINT2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Mouse | |
EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2 | |
HINT2 | |
IgG1 κ | |
Protein A purified |
5B10 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
84681 | |
Recombinant human HINT2 (18-163aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.