Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HINT2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550226

Catalog No. PA550226


Add to cart

Description

Description

The antibody detects endogenous levels of total HINT2 protein.

Histidine triad proteins, such as HINT2, are nucleotide hydrolases and transferases that act on the alpha-phosphate of ribonucleotides (Brenner, 2002 [PubMed 12119013]).[supplied by OMIM, Mar 2008]
Specifications

Specifications

HINT2
Polyclonal
Unconjugated
HINT2
Liquid
HINT2
Fusion protein corresponding to a region derived from internal residues of human histidine triad nucleotide binding protein 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
2.3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9BX68, Q9D0S9
HIT-17
Rabbit
IgG
100 μL
-20°C
Primary
68917, 84681
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit