Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4 beta 7/LPAM-1 Human anti-Human, Alexa Fluor 350, Clone: Hu117, R&D Systems
Human Monoclonal Antibody
Manufacturer: R&D Systems FAB10078U100UG
This item is not returnable.
View return policy
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal antibody specifically detects Integrin alpha 4 beta 7/LPAM-1 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 4 beta 7/LPAM-1 | |
Hu117 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Conjugated, Purified, Supernatant | |
Human | |
IgG1 | |
100 μg | |
Primary | |
3676 | |
Human |
Flow Cytometry | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49d/beta7, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), LPAM1, LPAM-1, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
Human Integrin alpha 4 beta 7/LPAM-1 | |
Protein A or G purified from cell culture supernatant | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
Monoclonal | |
Detects human Integrin alpha 4 beta 7/LPAM-1 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Vedolizumab. This product is for research use only. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title