Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Tocris Bioscience™ HIP-A
Potent, non-competitive EAAT inhibitor
Manufacturer:  Tocris Bioscience™ 2217/10
Catalog No. 22-171-0
This item is not returnable. View return policy
Description
Description
Potent, non-competitive excitatory amino acid transporter (EAAT) blocker. Preferentially inhibits glutamate-induced [3H]D-aspartate release (IC50 = 1.6 µM) rather than [3H]L-glutamate uptake (IC50 = 18 µM). Moderately selective; displays no affinity for NMDA and metabotropic glutamate receptors, and low affinity for AMPA and kainate receptors (IC50 values are 43 and 8 µM respectively). .
Specifications
Specifications
(±)-HIP-A
>98
10mg
(±)-HIP-A
172.14