Learn More
Tocris Bioscience™ HIP-B Available on GSA/VA Contract for Federal Government customers only.
Potent, non-competitive EAAT inhibitor
Manufacturer:  Tocris Bioscience™ 2218/10
Catalog No. 22-181-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent, non-competitive excitatory amino acid transporter (EAAT) blocker. Preferentially inhibits glutamate-induced [3H]D-aspartate release (IC50 = 1.2 µM) rather than [3H]L-glutamate uptake (IC50 = 16.9 µM). Moderately selective; displays no affinity for NMDA and metabotropic glutamate receptors, and low affinity for AMPA and kainate receptors (IC50 values are 35 and 45 µM respectively).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
(±)-HIP-B
>98
10mg
(±)-HIP-B
172.14