Learn More
Enzo Life SciencesSupplier Diversity Partner Hip polyclonal antibody (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPA766F
Host: Rabbit. Immunogen: Recombinant rat Hip. UniProt: P50503. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Bovine. Applications: IHC, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-512