Learn More
HIP14/ZDHHC17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154651AP
Catalog No. 50-173-0250
Add to cart
Description
Description
Palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. May be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. Has transforming activity. Mediates Mg(2+) transport.
Specifications
Specifications
HIP14/ZDHHC17
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E9PTT0, Q80TN5, Q8IUH5
DHHC 17, HIP 14, HIP 3, HIP14, HIP3, HSPC294, Huntingtin yeast partner H, HYPH, KIAA0946, Palmitoyltransferase ZDHHC17, ZDHHC17
Rabbit
IgG
150 μL
-20°C
Primary
23390, 320150, 366889
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZDHHC17
Liquid
Zdhhc17
HIP14; ZDHHC17 Fusion Protein Ag7763
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat