Learn More
HIP1R/HIP12 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304805AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.Specifications
HIP1R/HIP12 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O75146 | |
HIP1R | |
Between 300 and 350 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HIP1R | |
HIP12, HIP3, ILWEQ, HIP-12, HIP1-related protein, huntingtin interacting protein 12, huntingtin-interacting protein 1-related protein, huntingtin-interacting protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9026 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.