Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK1 Mouse anti-Mouse, Clone: 1-6, Active Motif™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$116.85 - $356.95

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen HIPK1
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2808 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39726
10 μg
Each for $116.85
Add to cart
 
50-199-2807 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39725
100 μg
Each for $356.95
Add to cart
 
Description

Description

The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.
Specifications

Specifications

Monoclonal
Primary
HIPK1
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit