Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIPK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514470
Description
This antibody is predicted to react with mouse based on sequence homology.
HIPK2 is a nuclear serine/threonine kinase that is linked to transcriptional regulation and apoptosis.
Specifications
HIPK2 | |
Polyclonal | |
Unconjugated | |
HIPK2 | |
1110014O20Rik; B230339E18Rik; cb1009; hHIPk2; HIPK2; homeodomain interacting protein kinase 2; homeodomain-interacting protein kinase 2; Nbak1; Nuclear body-associated kinase 1; PRO0593; si:dkey-16j24.2; si:dkey-239k7.1; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; wu:fk56d12 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
28996 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9H2X6 | |
HIPK2 | |
KLH conjugated synthetic peptide between 437-468 amino acids from the center region of human HIPK2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction