Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIPK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599799
Description
Antibody detects endogenous levels of total HIPK3.
HIPK3 is a serine/threonine kinase involved in signal transduction, cellular communication, and multidrug resistance of tumors and plays a role in the phosphorylation of FADD. HIPK3 negatively regulates apoptosis by promoting FADD phosphorylation. This kinase enhances androgen receptor-mediated transcription, and may act as a transcriptional corepressor for NK homeodomain transcription factors.
Specifications
HIPK3 | |
Polyclonal | |
Unconjugated | |
HIPK3 | |
Androgen receptor-interacting nuclear protein kinase; ANPK; DYRK6; Fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; HIPK3; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; mir-1902; nbak3; Nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; RP1-8L15.1; YAK1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10114 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H422 | |
HIPK3 | |
A synthesized peptide derived from human HIPK3(Accession Q9H422), corresponding to amino acid residues P930-V980. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction