Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hippocalcin like 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303318100UL
Description
Hippocalcin like 4 Polyclonal specifically detects Hippocalcin like 4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Hippocalcin like 4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
hippocalcin like 4, hippocalcin-like protein 4, HLP4DKFZp761G122 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-191 of human Hippocalcin like 4 (NP_057341.1). | |
100 μL | |
Vision | |
51440 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction