Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hippocalcin like 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Hippocalcin like 4
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093971
View Documents
Novus Biologicals
NBP303318100UL
100 μg Each for $320.71
Add to cart
 
NB093972
View Documents
Novus Biologicals
NBP30331820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Hippocalcin like 4 Polyclonal antibody specifically detects Hippocalcin like 4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Hippocalcin like 4
Unconjugated
Rabbit
Vision
PBS with 50% glycerol, pH7.3.
51440
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse, Rat
hippocalcin like 4, hippocalcin-like protein 4, HLP4DKFZp761G122
Recombinant fusion protein containing a sequence corresponding to amino acids 1-191 of human Hippocalcin like 4 (NP_057341.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit