Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hippuric Acid 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
495-69-2
C9H9NO3
179.17
MFCD00002692
QIAFMBKCNZACKA-UHFFFAOYSA-N
hippuric acid,n-benzoylglycine,glycine, n-benzoyl,benzoylglycine,benzamidoacetic acid,benzoylaminoacetic acid,hippurate,hippursaeure,phenylcarbonylaminoacetic acid,benzamidoessigsaeure
464
CHEBI:18089
2-benzamidoacetic acid
C1=CC=C(C=C1)C(=O)NCC(=O)O
495-69-2
C9H9NO3
179.17
MFCD00002692
QIAFMBKCNZACKA-UHFFFAOYSA-N
hippuric acid,n-benzoylglycine,glycine, n-benzoyl,benzoylglycine,benzamidoacetic acid,benzoylaminoacetic acid,hippurate,hippursaeure,phenylcarbonylaminoacetic acid,benzamidoessigsaeure
464
CHEBI:18089
2-benzamidoacetic acid
C1=CC=C(C=C1)C(=O)NCC(=O)O
Color White
Infrared Spectrum Authentic
Loss on Drying 1% max. (105°C)
Packaging Plastic bottle
Melting Point 188.0°C to 191.0°C
Quantity 500g
Assay Percent Range 98%
Linear Formula C6H5CONHCH2CO2H
Beilstein 09,225
Fieser 01,432
Merck Index 15,4753
Solubility Information Solubility in water: 1g/250 mL cold water. Other solubilities: freely soluble in hot water,1g/100mL chloroform- 1g/400mL ether,1g/60mL amyl alcohol,freely soluble in hot ether,pract.insoluble in benzene,cs2 and petr.ether
Formula Weight 179.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Hippuric acid, 98%
Show More Show Less

EINECSNumber : 207-806-3

RTECSNumber : MR8150000

TSCA : TSCA