Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

GE Healthcare HiPrep™ 26/10 Desalting Columns: PROMO

Provide reliable and reproducible desalting and buffer exchange with sample sizes of up to 15mL per column. GE Healthcare HiPrep 26/10 Desalting Column is a convenient, easy-to-use column prepacked 53mL column with Sephadex G-25 Fine for desalting and buffer exchange.

Manufacturer:  GE Healthcare 17508702PM

Catalog No. 45-000-267LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Saves time and labor as replacement for dialysis
  • Two to four columns can be used in series for sample sizes 30 to 60mL
  • Compatible with a laboratory pump or a chromatography system
  • Fittings are 1/16 in. for easy connection to AKTAdesign systems
  • Long-term and working pH range 2 to 13
  • Maximum flowrate: 15mL/min.

Specifications

Specifications

4 Pack
No
1.02 in.
4° to 30°C
Laboratory pump or chromatography system such as AKTA design
>20μm (Dry), 17 to 132μm (wet)
2.6cm
53mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit