GE Healthcare HiPrep™ Sephacryl™ S-300 HR Columns Promotion Available on GSA/VA Contract for Federal Government customers only.

Maximum recovery and minimum nonspecific adsorption

Manufacturer: GE Healthcare 17116701PM

 View more versions of this product

Catalog No. 45-000-197PM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Height (Metric) 60cm
    For Use With (Application) Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
    Height (English) 23.62 in.
    Material Polypropylene
    Max. Pressure 22psi
    Particle Size 25 to 75μm
    pH Range 3 to 11
    Sample Type Antibodies, serum proteins and mid-size proteins
    Quantity Each
    Storage Requirements 4° to 30°C
    Flow Rate <1.3mL/min.
    For Use With (Equipment) Low to medium pressure AKTA™ systems
    Bed Volume 120mL
    Diameter (Metric) Inner 1.6cm
    Support Type Allyl Dextran and N, N∽-methylenebisacrylamide
    Product Type HiPrep Sephacryl S-300 HR Column

    • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
    • Recommended linear flowrate 5 to 15cm/hr.
    • Designed for purifying antibodies, serum proteins and midsize proteins

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit