Learn More
Aviva Systems Biology Corp HIRIP3 Antibody - middle region (ARP58286_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIRIP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIRIP3. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-014-62278