Learn More
GE Healthcare His GraviTrap™ Columns PROMO
Use for purification of histidine-tagged proteins using gravity flow. His GraviTrap™ Columns enable fast and simple purification.
Manufacturer:  GE Healthcare 29361658
Catalog No. 45-000-006PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Prepacked with Ni Sepharose 6 Fast Flow media and ready to use
  • Allows direct purification of both clarified and unclarified cell lysates
  • Protein binding capacity approximately 40mg histidine-tagged protein/column
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives
Specifications
Specifications
1 mL
15mm
Polypropylene
45 to 165μm
10 x 1mL
6% Cross-linked Agarose
0.59 in.
His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification
3 to 12
His GraviTrap
4° to 30°C