Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ His Tag Monoclonal Antibody
Mouse Monoclonal Antibody
$127.00 - $999.00
Specifications
Antigen | His Tag |
---|---|
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Lyophilized |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
70-796-4MI
|
MilliporeSigma™
707964 |
Unconjugated | 3 μg |
Each for $127.00
|
|
||||
70-796-3MI
|
MilliporeSigma™
707963 |
Unconjugated | 100 μg |
Each for $999.00
|
|
||||
Description
The His Tag™ Monoclonal Antibody is a mouse monoclonal antibody (IgG1) directed against the His Tag sequence encoded by many of the Novagen™ expression vectors as well as many other commercially available vectors. The antibody recognizes five consecutive histidine residues regardless of the surrounding amino acid context. The high affinity (Kd = 5 x 10-8 to 1 x 10-9 M) enables sensitive, specific detection of His Tag fusion proteins at antibody concentrations of 0.1-0.2μg/ml. The His Tag Monoclonal Antibody binds to N-terminal, C-terminal, and internal His Tag sequences. This antibody also detects the recombinant marker bands in the Perfect Protein™ and Trail Mix™ Western Markers for convenient visualization of accurate internal standards on Western blots*. For lowest background in Western blotting applications, Alkali-soluble Casein is recommended as a blocking agent. The 100μg package size provides enough purified antibody for 1000 ml of working solution.Specifications
His Tag | |
Monoclonal | |
Lyophilized | |
RUO | |
IgG1 | |
-20°C |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized, BSA-free | |
Primary | |
HisHisHisHisHis, N-terminal, C-terminal or internal |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title