Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Mouse anti-Human, Non-species specific, Sheep, Alexa Fluor™ 750, Clone: HIS.H8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP231055AF750

Catalog No. NP2310A70


Add to cart

Description

Description

His Tag Monoclonal antibody specifically detects His Tag in Human, Non-species specific, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

His Tag
HIS.H8
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
Mouse
IgG2b
0.1 mL
Primary
Human, Nonspecies specific, Sheep
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
6x His synthetic peptide.
Protein A purified
Store at 4°C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit