Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Mouse, HRP, Clone: 6G2A9, THE™, GenScript™

Catalog No. 502120474
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0474 Supplier Genscript Corporation Supplier No. A00612
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water to the final concentration of 0.5 mg/mL. GenScript THE™ His Tag Antibody [HRP], mAb, Mouse is HRP conjugated THE™ Anti-His Monoclonal Antibody (Mouse) (A00186) purified from mice ascites by protein A affinity column. This product is suitable for detecting the expression level of His fusion proteins or His by various immunoassays.

His Tag Monoclonal antibody specifically detects His Tag in Tag samples. It is validated for ELISA, Western Blot

Specifications

Antigen His Tag
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6G2A9
Conjugate HRP
Formulation PBS with 1% BSA and 0.01% thimerosal; pH 7.4
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Mouse
Immunogen A synthetic peptide HHHHHH coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.